Learn More
AdipoGen 2-Tosyloxycyclohexanone
SDP
Supplier:  AdipoGen CDXT0131G001
Encompass_Preferred
Chemical. CAS 81447-34-9. Formula C13H16O4S. MW 268.3. Synthetic. Alpha-​tosyloxy ketone used for synthesis.
Catalog No. 50-255-9988