Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 2-Tributylstannylpyridine, 17997-47-6, MFCD00052052, 5g

Supplier:  Matrix Scientific 0043515G

Encompass

Molecular Formula C17H31NSn, Purity 90%, Molecular Weight 368.13, Melting Point 132-135°, Boiling Point 130-132/0.8 Torr

Catalog No. 50-147-3088


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.