Learn More
Medchemexpress LLC 2-(Trifluoromethyl)cinnamic acid | 2062-25-1 | 99.8% | C10H7F3O2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY75424A
Encompass_Preferred
2-(Trifluoromethyl)cinnamic acid is a cinnamic acid derivative that inhibits the proton pump (H+/K+-ATPase), thereby reducing gastric acid secretion. It also improves delayed gastric emptying and can be used in research on gastric diseases such as acute gastritis and gastric ulcers.
  • Cinnamic acid derivative
  • Inhibits proton pump (H+/K+-ATPase)
  • Reduces gastric acid secretion
  • Improves delayed gastric emptying
  • Used in research on gastric diseases such as acute gastritis and gastric ulcers
Catalog No. 50-003-58512