Learn More
Medchemexpress LLC 2-Undecanol 10mM 1mL | 1653-30-1 | 172.31 | C11H24O | 1 ML

Supplier: Medchemexpress LLC HY11568410MM 1ML
2-Undecanol is a research-grade secondary alcohol supplied as either the neat material or as a 10 mM solution in DMSO (1 mL). It is provided for laboratory research and analytical applications, including studies of volatile compounds and pheromone components. Intended for research use only.
- Research-grade secondary alcohol.
- Available as 10 mM solution in DMSO, 1 mL, and in multiple bulk sizes.
- Purity 98.0%.
- Molecular formula C11H24O; molecular weight 172.31.
- Suitable for analytical and volatile compound research.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.