Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Undecanol 1g | 1653-30-1 | 1 G
SDP

Supplier:  Medchemexpress LLC HY1156841G

Encompass_Preferred

2-Undecanol (undecan-2-ol) is a C11 aliphatic secondary alcohol used as a research reagent and reference standard for studies of volatile organic compounds, pheromone components, and flavor/fragrance chemistry.

  • Cas number: 1653-30-1.
  • Molecular formula C11H24O; molecular weight 172.31 g/mol.
  • Purity 98.0%.
  • Appearance liquid, colorless to light yellow; density ~0.827 g/cm3.
  • Recommended storage: 4°C sealed; in solvent store at -80°C up to 6 months.
  • Pack size: 1 G (liquid).

Catalog No. 50-002-26516


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.