Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-undecyl-4R-thiazolidinecarboxylic acid | 298186-80-8 | 98.0% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY11940110MG

Encompass_Preferred

A selective small-molecule antagonist of the sphingosine-1-phosphate receptor 3 (S1P3) used in research to probe S1P3-mediated signaling and intracellular calcium mobilization.

  • S1P3 receptor antagonist used for in vitro pharmacology studies.
  • Chemical formula C15H29NO2S.
  • Molecular weight 287.5 g/mol.
  • Purity 98.0% by HPLC.
  • White to off-white solid appearance.
  • Typical quantity: 10 mg vials; store powder at -20°C for long-term stability.

Catalog No. 50-002-34354


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.