Learn More
Medchemexpress LLC 2-(undecyloxy)ethanol | 38471-47-5 | MFCD06252492 | 99.1% | 216.37 | C13H28O2 | 500 MG

Supplier: Medchemexpress LLC HYW040313500MG
2-(Undecyloxy)ethanol is a research-grade pheromone supplied as a reagent for laboratory studies. It is reported to be produced by male Monochamus alternatus and is provided with analytical information for research use.
- chemical identity: 2-(undecyloxy)ethanol, CAS 38471-47-5
- high purity suitable for research use
- molecular formula C13H28O2 and molecular weight ~216.37 g/mol
- available in multiple small-scale lab sizes for experimental work
- typical use: pheromone studies and analytical research
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.