Learn More
Zebra Technologies Corporation Label, Kimdura Polypropylene, 3x2in (76.2x50.8mm); TT, PolyPro 4000T, High Performance Coated, Permanent Adhesive, 3in (76.2mm) core, 2230/roll, 4/box, Plain
Supplier:  Zebra Technologies Corporation 10011689
Encompass_Preferred
This item has a minimum order quantity of 4 per supplier requirements
Label, Kimdura Polypropylene, 3x2in (76.2x50.8mm); TT, PolyPro 4000T, High Performance Coated, Permanent Adhesive, 3in (76.2mm) core, 2230/roll, 4/box, Plain
Catalog No. NC2123766