Learn More
Avanti Polar Lipids 20:0 Lyso PC | 108341-80-6 | MFCD09841690 | 25mg
Supplier:  Avanti Polar Lipids 855777P25MG
Encompass
20:0 Lyso PC | Purity: >99% LPC; may contain up to 10% of the 2-LPC isomer | MW: 551.740 | 108341-80-6 | MFCD09841690 | 25mg
Catalog No. NC1624974