Learn More
MSC Pelican Products Clamshell Hard Case, Layered Foam, IP67 Rated, Black
Supplier:  MSC 84189620
Encompass
This Pelican Products Clamshell Hard Case is a medium-sized protective transport case featuring layered foam for equipment protection. It is designed with a solid wall, open cell core, and an O-ring seal to ensure durability and protection against crushing, dust, and water. The case also includes an automatic pressure equalization valve and is IP67 rated, making it suitable for demanding environments.
  • Solid wall design
  • Crush proof
  • O-ring seal
  • Automatic pressure equalization valve
  • Lightweight
  • Watertight
  • Open cell core
  • Dust proof
  • Strong
  • Includes foam
  • Padlockable
  • IP67 standards
Catalog No. 50-005-31209