Learn More
Cayman Chemical 20-Deoxyngenol 10mg
SDP
Supplier:  Cayman Chemical 3322510 MG
Encompass_Preferred
A diterpenoid; inhibits oxidative stress-induced decreases in autophagic flux and apoptosis in isolated mouse chondrocytes at 10 mM; reduces bone erosion and cartilage calcification in joints in a mouse model of injury-induced osteoarthritis at 20 mg/kg
Catalog No. 50-294-7233