Learn More
Medchemexpress LLC 20α-dihydroprogesterone | 145-14-2 | MFCD00009823 | 97.1% | 316.5 g/mol | C21H32O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1130625MG
Encompass_Preferred
20α-dihydroprogesterone is an endogenous metabolite of progesterone supplied as a research-grade powder for laboratory use. It is used as an analytical reference and in biochemical or pharmacological studies. Chemical identity: C21H32O2, molecular weight 316.5 g/mol, CAS 145-14-2. Typical purity is about 97.1%, and the product is commonly supplied in 5 mg amounts. Store and handle per supplier documentation.
Catalog No. 50-002-64194