Learn More
Medchemexpress LLC 20-HC-Me-Pyrrolidine | 99.7% | C28H47NO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1450011MG
Encompass_Preferred
20-HC-Me-Pyrrolidine (compound AI-3d) is a potent Aster protein inhibitor with IC50s of 0.11 μM, 0.06 μM, and 0.71 μM for Aster-A, Aster-B, and Aster-C, respectively. It blocks the ability of Asters to bind and transfer cholesterol and inhibits the movement of low-density lipoprotein (LDL) cholesterol to the endoplasmic reticulum (ER).
  • Potent aster protein inhibitor.
  • Blocks the ability of asters to bind and transfer cholesterol.
  • Inhibits the movement of low-density lipoprotein (LDL) cholesterol to the endoplasmic reticulum (ER).
  • For research use only.
  • White to off-white solid appearance.
  • Purity of 99.7%.
Catalog No. 50-003-54850