Learn More
Medchemexpress LLC 20-hede | 240427-90-1 | 97.1% | 324.50 | C20H36O3 | 1 MG

Supplier: Medchemexpress LLC HY1015271MG
20-HEDE (WIT 002) is a small-molecule antagonist of 20-hydroxyeicosatetraenoic acid (20-HETE) provided as a high-purity research chemical for laboratory studies of cytochrome P450 metabolites and eicosanoid signaling. It is intended for research use only and includes documented solubility and storage recommendations to support experimental handling.
- High purity suitable for research applications.
- Molecular weight 324.50; molecular formula C20H36O3.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
- Storage: pure form -20°C up to 3 years; in solvent -80°C 6 months or -20°C 1 month.
- Available in small-quantity packaging for experimental use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.