Learn More
Cayman Chemical 20-hydroxy Ecdyson 10mg
SDP
Supplier:  Cayman Chemical 1614510 MG
Encompass_Preferred
An ecdysteroid hormone produced in arthropod species that induces developmental changes associated with ecdysis and the completion of metamorphosis; used as a tool in inducible gene regulation systems controlled by reporter cells transfected with the ecdysone receptor
Catalog No. 50-297-1640