Learn More
Medchemexpress LLC 20-Hydroxyecdysone | 5289-74-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN69795MG
Encompass_Preferred
Crustecdysone (20-Hydroxyecdysone) is a naturally occurring ecdysteroid hormone isolated from *Serratula coronata*. It controls the ecdysis (moulting) and metamorphosis of arthropods, inhibits caspase activity, and induces autophagy via the 20E nuclear receptor complex, EcR-USP. It also exhibits regulatory or protective roles in the cardiovascular system and is an active metabolite of Ecdysone.
  • Naturally occurring ecdysteroid hormone.
  • Controls ecdysis (molting) and metamorphosis in arthropods.
  • Inhibits caspase activity.
  • Induces autophagy via the 20E nuclear receptor complex, EcR-USP.
  • Exhibits regulatory or protective roles in the cardiovascular system.
  • Induces cell autophagy and caspase-3 activity in the remodeling fat body of Drosophila.
  • Reduces blood pressure and prevents dilated cardiac hypertrophy in SHR rats.
Catalog No. 50-004-18246