Learn More
Cayman Chemical 20 R-ProtopnaxatrIol 10mg
SDP
Supplier:  Cayman Chemical 3653010 MG
Encompass_Preferred
A degradation product of panaxatriols; cytotoxic to P388, L1210, and K562 leukemia cells (EC50s = 34, 32, and 38 µM); inhibits EGFR (IC50 = 24.1 µM) in a cell-free assay; cytotoxic to HepG2 cells from 20-100 µM
Catalog No. 50-291-2199