Learn More
Medchemexpress LLC HY-12316 5mg Medchemexpress, 20(S)-Hydroxycholesterol CAS:516-72-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123165MG
Encompass_Preferred
Medchemexpress, HY-12316 5mg 20(S)-Hydroxycholesterol CAS:516-72-3 20(S)-hydroxyCholesterol (20α-Hydroxycholesterol) is an allosteric activator of the oncoprotein smoothened (Smo) that activates the hedgehog (Hh) signaling pathway with an EC 50 of 3 μM in a gene transcription reporter assay using NIH3T3 cells [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2576