Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 20-(tert-butoxy)-20-oxoicosanoic acid | 683239-16-9 | MFCD23136041 | 99.9% | 398.62 g/mol | C24H46O4 | 25 G
SDP

Supplier:  Medchemexpress LLC HYW03459725G

Encompass_Preferred

20-(tert-butoxy)-20-oxoicosanoic acid is an alkyl chain linker used as a non-cleavable linker for antibody-drug conjugates (ADCs) and as an alkyl chain-based linker in PROTAC synthesis. Supplied as a high-purity research-grade compound, it is intended for use in medicinal chemistry and synthetic linker construction.

  • High purity (99.9%).
  • Molecular weight 398.62 g/mol.
  • Chemical formula C24H46O4.
  • Available in 25 g packaging.
  • Powder storage: -20°C for long-term stability.
  • Intended application: ADC and PROTAC linker synthesis.

Catalog No. 50-002-03074


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.