Learn More
Biocare Medical, LLC Tween 20 - 500 ml
SDP
Supplier:  Biocare Medical, LLC TWN20M
Encompass_Preferred
Tween 20 is a detergent (surfactant) commonly added to buffers and reagents for immunohistochemistry. Its purpose is to decrease background staining and enhance reagent spreading in automated and manual procedures. It can also be used on an automated staining system.
Catalog No. 50-832-99