Learn More
Chemglass Life Sciences Chemglass 200 μL Automation Clear Tips for Tecan, Sterile, Blister Pack (96 x 2/pack; 2,304/case)
SDP
Supplier:  Chemglass Life Sciences CLS3499A103
Encompass_Preferred
Chemglass 200 μL automation clear tips are sterile, medical-grade polypropylene tips engineered for high-throughput automated liquid handling on Tecan platforms. They are blister-packed in nests to reduce contamination and supplied 2 x 96 per blister with 2,304 tips per case.
  • 200 μL capacity.
  • Sterile, electron beam sterilized.
  • Medical-grade polypropylene construction.
  • DNase/RNase free.
  • Blister-packed nest format for automation.
  • 2,304 tips per case (96 x 2 per blister pack).
Catalog No. 50-273-3245