Learn More
Chemglass Life Sciences Chemglass 200 μL automation tips for Tecan, clear, sterile, boxed (96/rack, 10 racks/pk)
SDP
Supplier:  Chemglass Life Sciences CLS3499A403
Encompass_Preferred
200 μL automation pipette tips designed for high-throughput liquid handling with Tecan systems. Sterile, ultra-clear, medical-grade polypropylene construction helps minimize contamination and supports accurate pipetting for genomics, proteomics, and routine laboratory workflows.
  • 200 μL nominal volume for standard microplate transfers.
  • Sterile, DNase/RNase-free, electron beam sterilized.
  • Medical-grade polypropylene for chemical compatibility and clarity.
  • Supplied nested: 96 tips per rack, 10 racks per pack, 50 racks per case.
  • Compatible with Tecan automated liquid handling platforms.
  • Designed for high-throughput workflows and reproducible results.
Catalog No. 50-273-3235