Learn More
Tecan 200L Tips ANSI NFil Ref
Supplier:  Tecan 30057814
Encompass_Preferred
Genuine Tecan 200 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). ANSI format, non-filtered, conductive. These tips are designed to fit into a Disposable Tip (DiTi) box as a refill. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 24 racks of 96-tips, 2,304 tips/sales unit. LiHa Disposable Tips have been carefully designed and validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality and reproducibility, ensuring reliable liquid handling performance. Manufacturing is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures and premium raw materials.
Catalog No. NC0527903