Learn More
Chemglass Life Sciences Clamp, Stainless Steel, Quick Release, 200mm Duran Flange
SDP
Supplier:  Chemglass Life Sciences CG14105
Encompass_Preferred
Clamp, Stainless Steel, Quick Release, 200mm Duran Flange. Made entirely from stainless steel, the clamp has three sections that firmly hold reaction vessel and lid together. Quick release mechanism has a screw adjustment for tension applied to the flange. 200mm Duran Flange.
Catalog No. CG14105