Learn More
Polycarbin 200 ul Pipette Tips, LTS Compatible, Refill Tower (Non-Sterile)
SDP
Supplier:  Polycarbin PC010506R
Encompass_Preferred
Polycarbin's LTS compatible refill system is designed to deliver an exceptional product that is less carbon and plastic-intensive. Refill tower housing is manufactured using post-consumer recycled content, and are shrink wrapped and packaged in an easy to recycle cardboard sleeve. Pipette tips do not contain recycled content.
Catalog No. NC3073513