Learn More
AUTOMATION TRAINER 200L Tecan MCA-Type 96-Format, Clear, Unfiltered, Sterile, Racked
SDP
Supplier:  AUTOMATION TRAINER TM200CLUSR
Encompass_Preferred
Automation Trainers Tecan MCA Type pipette tips are designed to be a direct replacement to Tecans 96-Format MCA tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition are not required.- Fits Tecan Liquid Handling (MCA) & Flexible-Channel (FCA) Arms - High-Grade Polypropylene Construction - 50 L & 200 L Max. Working Volumes - 50 L (Case of 50 96-Tip Racks), 200 L (Case of 50 96-Tip Racks) - Packaging: Case of Individually-Wrapped Racks
Catalog No. NC2219051