Learn More
Chemglass Life Sciences Pipette Tips, 200 μL, Clear, Bulk Packed (1,000/Pack, 10,000/Case)
SDP
Supplier:  Chemglass Life Sciences CLS3499E200
Encompass_Preferred
Clear, non-sterile 200 μL pipette tips for routine laboratory pipetting. Bulk-packed in nest format for high-volume workflows; universal fit for most single- and multichannel pipettors.
  • Bulk packed: 1,000 tips per pack, 10,000 tips per case.
  • Nominal volume: 200 μL.
  • Ultra-clear polymer for visual accuracy and solvent resistance.
  • Non-sterile, intended for routine laboratory use.
  • Universal fit for most single- and multichannel pipettors.
  • Low-retention, fine-point design reduces sample adhesion.
  • Certified RNase-, DNase-, human DNA-, and pyrogen-free.
Catalog No. 50-273-3208