Learn More
USA Scientific Inc 200 ul TipOne natural, beveled tip, racks, sterile, case of 8 packs
Supplier:  USA Scientific Inc 11111880
Encompass
TipOne 200 ul natural, beveled pipet tips in racks, sterile, 80 racks of 96 tips (7680 tips)
Catalog No. NC1610533