Learn More
MSC Value Collection ZMSCV70201 3/16" ID x 7/16" OD Dash 201 Viton O-Ring 1/8" Thick, Round Cross Section, Shore 65 - 75, -15 to 400°F
Supplier:  MSC 02050789
Encompass
Value Collection ZMSCV70201 3/16" ID x 7/16" OD Dash 201 Viton O-Ring 1/8" Thick, Round Cross Section, Shore 65 - 75, -15 to 400°F
Catalog No. NC1466995