Learn More
Refrigiwear Inc. 2011 REG HIGH VIS LIME LARGE KNIT GLOVE WITH DOT GRIP AND TOUCH SCREEN
SDP
Supplier:  Refrigiwear Inc. 2011RHVLLAR
Encompass_Preferred
The functional Touchscreen Double Dot Glove is constructed to make you unstoppable on the job. With a HiVis lime PVC dot grip pattern giving you superior gripping power & keeping you seen in low light conditions, you can feel confident products are secure in your hands. Double-sided PVC dots are added to extend the wear of these gloves, so you don’t need to worry about frequent replacement. Touchscreen capability on three fingers adds convenience to daily life on the job by limiting the need to remove handwear to operate touchscreen devices & ensuring your handwear is constantly keeping you protected in cold conditions. Since the Touchscreen Double Dot Glove is sold by the dozen, you can make sure there’s a pair for you & any member of your warehouse selector team. Touchscreen capability on index, thumb & middle fingers Brushed acrylic loop terry Double-sided for extended wear HiVis lime PVC dot grip pattern Sold by the dozen / Volume discounts available  
Catalog No. NC3867132