Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Nucleocapsid Detection ELISA Kit
Supplier:  Sino Biological KIT405881
Encompass_Preferred
The kit has been verified by high purity SARS-CoV-2 (2019-nCoV) Nucleocapsid Protein (Cat# 40588-V08B). It can recognize recombinant SARS-CoV-2 variant Proteins, and SARS-CoV Nucleocapsid Protein, but no react with recombinant MERS-CoV, HCoVs Nucleocapsid protein and Influenza Nucleoprotein.
Catalog No. NC1792337