Learn More
Cayman Chemical 20a-dIhydro PrednIsolon 5mg
SDP
Supplier:  Cayman Chemical 265025 MG
Encompass_Preferred
A metabolite of prednisolone; has been found in the urine of cattle following ACTH administration but not in naturally stressed or non-stressed cattle
Catalog No. 50-293-4571