Learn More
Alkali Scientific 20mL Clear Glass Storage Vial with White Closed Top Cap, 100/Pk
SDP
Supplier:  Alkali Scientific SCNT020STORAGE
Encompass_Preferred
The Alkali Scientific™ 20 mL Clear Glass Vial is designed for reliable sample storage and handling in laboratory and analytical applications. Made from high quality clear glass, the vial provides excellent chemical resistance and full visibility of contents. Each vial comes pre assembled with a 22 400 white closed top polypropylene (PP) cap and 1.5 mm thick silver aluminum foil/PE liner septa, ensuring a secure, leak free seal and chemical inertness. Supplied with dividers to prevent contact and breakage during storage and transport, this pack of 100 vials is ideal for chromatography, sample storage, and high throughput laboratory workflows. Key Features 20 mL capacity for reliable sample storage and handling Made from high quality clear glass for excellent chemical resistance and visibility Pre assembled 22 400 white closed top PP cap with 1.5 mm thick silver aluminum foil/PE liner septa Secure, leak free seal with chemical inertness for analytical application
Catalog No. 50-006-62942