Learn More
Teknova 20mM HEPES, 150mM Sodium Chloride, pH 7.0. 1000mL. Endotoxin Tested, Sterile. 8 pack.
SDP
Supplier:  Teknova H63358PK
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-274-5954