Learn More
Ace Glass, Inc. 20MM POR D FILTER FUNEL 20MM POR D FILTER FUNEL
SDP
Supplier:  Ace Glass, Inc. 718708
Encompass_Preferred
20mm porosity D (10-20 micron) filter funnel with a 60 degree angle. Funnel is useful where it is necessary to wash the precipitation and re-dissolve with chemicals that would attach filter paper.
Catalog No. 50-974-303