Learn More
Chemglass Life Sciences 20mm Quartz Fritted Disc, Extra Coarse
SDP
Supplier:  Chemglass Life Sciences CGQ020709
Encompass_Preferred
20mm Quartz Fritted Disc, Extra Coarse. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020709