Learn More
Cayman Chemical 20OAcetylngenol3ngelate 10mg
SDP
Supplier:  Cayman Chemical 3990410 MG
Encompass_Preferred
A diterpene ester with anticancer activity; inhibits the growth of K562, HL-60, KT-1, MCF-7/adr, A549, and NIH3T3 cells at 12.5 µM; induces cell cycle arrest at the G2/M phase, apoptosis, and necrosis in K562 cells from 31.25-500 nM
Catalog No. 50-294-2508