Learn More
Medchemexpress LLC (20R)-Ginsenoside Rg3 | 38243-03-7 | 785.01 | 1 ML
Supplier: Medchemexpress LLC HYN137610MM/1ML
(20R)-ginsenoside Rg3, also known as (20R)-Propanaxadiol, is one of the active compounds found in ginseng root. It is recognized for its ability to inhibit angiogenesis and possesses anti-carcinogenic and antimetastatic effects.
- Inhibits the proliferation of human umbilical vein endothelial cells (HUVEC) with an IC50 of 10 nM.
- Dose-dependently suppresses capillary tube formation on Matrigel.
- Significantly attenuates VEGF-induced chemoinvasion of HUVEC and ex vivo microvascular sprouting.
- Remarkably abolishes bFGF-induced angiogenesis in an in vivo Matrigel plug assay.
- Shows anti-fatigue effects in mice via intranasal administration.