Learn More
Selleck Chemical LLC (20S)-Protopanaxatriol S3617-5mg
SDP
Supplier:  Selleck Chemical LLC S36175MG
Encompass_Preferred
(20S)-Protopanaxatriol (g-PPT 20(S)-APPT) a neuroprotective metabolite of ginsenoside protopanaxatriol (g-PPT) is a functional ligand for both GR and ERbeta and could modulate endothelial cell functions through the glucocorticoid receptor (GR) and estrogen receptor (ER)
Catalog No. 50-310-9332