Learn More
TARGETMOL CHEMICALS INC 20S-PROTOPANAXATRIOL 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T281025MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (20S)-Protopanaxatriol (g-PPT)(g-PPT) a metabolite of ginsenoside could regulate endothelial cell functions through the estrogen receptor and glucocorticoid receptor. purity: 99%
Catalog No. 50-258-0550