Learn More
Pacer Global LLC 20μL Pipette Tips, Low Retention, Filter, Sterile, 96 tips per rack, 10 racks per pack
SDP
Supplier:  Pacer Global LLC MLSPTFS20J/DUP
Encompass_Preferred
20μL Universal Pipette Tips, Low Retention, Filter, Sterile, 96 tips/rack, 10 racks/pack. Lot traceable and DNase free, RNase free, and non-pyrogenic. Additional info on MLS-PTFS-20J: Sterile & Super low retention. Made of clear, high-quality polypropylene. Molded volume graduation markings. Interior support ribs prevent rack from warping. Smooth, hydrophobic surfaces to ensure accuracy. Lids have a stacking feature for stability and a secure locking tab. Packaged in yellow rigid autoclavable racks. Filters reduce cross contamination and prevent damaging aerosols from penetrating into the pipette. Certifications:ISO11137ISO13485:2016ISO9001:2015 These pipette tips are universal fit and are compatible with major pipettes, including Eppendorf and Corning. Further discounted pricing available for spot buys of minimum 200 cases.
Catalog No. NC1915693
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.