Learn More
Bellco Glass, Inc. 20µL Pipette, MLA, Precision, Black
SDP
Supplier:  Bellco Glass, Inc. 451900002
Encompass_Preferred
20µL Pipette, MLA, Precision, BlackTHE CLASSIC MODELSingle-stroke “to deliver” operationAvailable in 28 fixed volumes ranging from 5 μL- 1000 μLEasily adjusted as needed to achieve accurate performance on dense or viscous liquids All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorBlackPrimary MaterialAluminum Alloy & Stainless SteelVolume20 μLVistaLab Tip Fit SizeSmall 250μLUniversal FitNoAccuracy±3.00%Precision1.35%
Catalog No. NC3771675