Learn More
Electron Microscopy Sciences Steinheil Magnifier, 20x
SDP
Supplier:  Electron Microscopy Sciences 68148
Encompass_Preferred
A biconvex lens. Pocket size. The sliding frame is designed for the storage of the lens after use. It is good for the preliminary inspection of microscopic specimens. It is lightweight and has excellent chromatic correction with a reasonably wide field of view. The magnifier helps the user eliminate bulky and heavier conventional magnifiers.
Magnification: 20x
Catalog No. 50-291-25