Learn More
Cayman Chemical 21-DeoxycortIson 10mg
SDP
Supplier:  Cayman Chemical 3523610 MG
Encompass_Preferred
A corticosteroid metabolite of 11-keto progesterone; levels are increased in patients with congenital adrenal hyperplasia
Catalog No. 50-290-9336