Learn More
Medchemexpress LLC 21-desacetyldeflazacort | 13649-57-5 | 99.2% | 399.48 g/mol | C23H29NO5 | 1 MG

Supplier: Medchemexpress LLC HY1000851MG
21-Desacetyldeflazacort is the active metabolite of the glucocorticoid prodrug deflazacort, supplied as a high-purity analytical standard for biochemical and pharmacological research. Provided as a solid, it is suitable for metabolite profiling, mechanism-of-action studies, and analytical method development.
- High purity (99.23%).
- Molecular weight 399.48 g/mol.
- Solid, white to off-white appearance.
- Powder stable at -20°C for up to 3 years.
- Suitable as a reference standard for metabolite and pharmacology studies.
- Supplied in a 1 mg quantity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.