Learn More
Medchemexpress LLC 21-desacetyldeflazacort-d5 | 13649-57-5 | 99.3% | 404.51 g/mol | C23H24D5NO5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY100085S1MG
Encompass_Preferred
21-Desacetyldeflazacort-d5 is the deuterium-labeled analogue of 21-desacetyldeflazacort provided as a research reference standard. It is intended for research use only and is characterized by its deuterated formula and high reported purity.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative analysis.
  • High stated purity (99.34%) for reliable analytical performance.
  • Accurately measured small-scale quantities suitable for method development.
  • Molecular weight approximately 404.51 g/mol for calculation and reporting.
  • Provided with CAS registry number and supporting documentation such as COA and SDS.
Catalog No. 50-002-15514