Learn More
Medchemexpress LLC 21-desacetyldeflazacort-d5 | 13649-57-5 | 99.3% | 404.51 g·mol⁻¹ | C23H24D5NO5 | 10 MG

Supplier: Medchemexpress LLC HY100085S10MG
21-Desacetyldeflazacort-d5 is a deuterium-labeled internal standard of 21-desacetyl deflazacort used for quantitative analytical assays such as LC-MS/MS. It is supplied for use as an internal standard in pharmacokinetic, metabolic, and bioanalytical studies where accurate quantitation of the metabolite is required.
- High isotopic purity for reliable internal standardization.
- Purity 99.3% (manufacturer reported) ensures minimal interference.
- Molecular formula C23H24D5NO5 and molecular weight 404.51 g·mol⁻¹.
- Available in small, pre-weighed quantities suitable for trace analysis.
- Provided with analytical data and safety documentation for assay validation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.