Learn More
Cayman Chemical 21hydroxy HenIcosnoIc ACD 10mg
SDP
Supplier:  Cayman Chemical 2465410 MG
Encompass_Preferred
A hydroxy fatty acid methyl ester; a substrate for lactonizing lipase from Pseudomonas nov. sp. 109
Catalog No. 50-301-5687