Learn More
Seneca Medical Inc 21IN IV EXT SET CS-50
Supplier:  Seneca Medical Inc 885350
Encompass_Preferred
IV ADMINISTRATION SETS. PERIPHERAL EXTENSION SETS. STANDARD BORE EXTENSION SETS. FEMALE LUER CONNECTOR, DISTAL SPIN-LOCK CONNECTOR. DEHP-FREE, LATEX-FREE. PRIMING VOLUME:3ML. LENGTH: 21 in. (53.3CM). 50 PER CASE.
Catalog No. NC9591259