Learn More
Sigma Aldrich Fine Chemicals Biosciences 22 Dithiodimethylenedifura1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W3146091KGK
Encompass
Difurfuryl disulfide is a sulfur-containing volatile compound mainly found in roasted coffee. It is also the main degradation product of furfuryl mercaptan under fenton-type reaction conditions.
Catalog No. 50-290-2374